Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Nicotinate, Spectrum™ Chemical
SDP

Catalog No. 18620730 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-730 500 g
18-620-729 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-730 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E2510500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Synonym: Ethyl Pyridine-3-Carboxylate, Ethylnicotinate, Ethyl Pyridine-3-Carboxylate

Chemical Identifiers

CAS 614-18-6

Specifications

Boiling Point 223°C to 224°C (433°F to 435°F)
Quantity 500 g
Molecular Weight (g/mol) 151.17
Percent Purity 99%
Grade Ungraded
Physical Form Liquid
Chemical Name or Material Ethyl Nicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.