Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl nipecotate

Catalog No. 19436025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-436-025G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-436-025G Supplier Sigma Aldrich Supplier No. 19436025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl nipecotate

Specifications

CAS 5006-62-2
Density 1.012 g/mL (at 25°C (literature))
Boiling Point 102°C to 104°C (7 mmHg)
Molecular Formula C8H15NO2
Refractive Index n20/D 1.460 (literature)
Linear Formula C8H15NO2
MDL Number MFCD00005991
Quantity 100 mg
Synonym 3-Piperidinecarboxylic acid ethyl ester; Ethyl 3-piperidinecarboxylate; NSC 158451
Molecular Weight (g/mol) 157.21
Percent Purity 96%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.