Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl nitroacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
626-35-7
C4H7NO4
133.1
FTKASJMIPSSXBP-UHFFFAOYSA-N
ethyl nitroacetate,nitroacetic acid ethyl ester,acetic acid, nitro-, ethyl ester,nitro ethyl acetate,ethylnitroacetate,acetic acid, 2-nitro-, ethyl ester,ethylnitroacetat,ethyinitroacetate,ethyl-2-nitroacetate,zlchem 269
69379
ethyl 2-nitroacetate
CCOC(=O)C[N+](=O)[O-]
626-35-7
C4H7NO4
133.1
FTKASJMIPSSXBP-UHFFFAOYSA-N
ethyl nitroacetate,nitroacetic acid ethyl ester,acetic acid, nitro-, ethyl ester,nitro ethyl acetate,ethylnitroacetate,acetic acid, 2-nitro-, ethyl ester,ethylnitroacetat,ethyinitroacetate,ethyl-2-nitroacetate,zlchem 269
69379
ethyl 2-nitroacetate
CCOC(=O)C[N+](=O)[O-]
Boiling Point 105.0°C to 107.0°C (25.0 mmHg)
Specific Gravity 1.199
Quantity 5g
Infrared Spectrum Authentic
Color Colorless to Yellow
Flash Point 92°C
Density 1.1990g/mL
Packaging Glass bottle
Refractive Index 1.4235 to 1.4255
Assay Percent Range 97.5% min. (GC)
Linear Formula O2NCH2CO2C2H5
Beilstein 02,225
Formula Weight 133.1
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Ethyl nitroacetate
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 210-944-7