Learn More
Medchemexpress LLC Ethyl octanoate | 106-32-1 | MFCD00009552 | 99.9% | 172.27 g/mol | C10H20O2 | 25 G
SDP
Catalog No. 5000303014
Encompass_Preferred
Ethyl octanoate is an ethyl ester of octanoic (caprylic) acid (C10H20O2, 172.27 g/mol) with a characteristic fruity aroma. It is provided as a high-purity reagent for biochemical assays, analytical standards, flavor and fragrance research, and organic synthesis, with defined storage and shelf-life recommendations.
  • High purity 99.86% suitable for analytical and assay applications.
  • Clear liquid with a fruity odor useful for sensory and flavor studies.
  • Molecular formula C10H20O2 and molecular weight 172.27 g/mol for calculation and reporting.
  • Stable at room temperature with extended shelf life; solvent storage options at -20°C or -80°C.
Catalog No. 50-003-03014 Supplier Medchemexpress LLC Supplier No. 25GHYW015305
Ethyl octanoate is an ethyl ester of octanoic (caprylic) acid (C10H20O2, 172.27 g/mol) with a characteristic fruity aroma. It is provided as a high-purity reagent for biochemical assays, analytical standards, flavor and fragrance research, and organic synthesis, with defined storage and shelf-life recommendations.
  • High purity 99.86% suitable for analytical and assay applications.
  • Clear liquid with a fruity odor useful for sensory and flavor studies.
  • Molecular formula C10H20O2 and molecular weight 172.27 g/mol for calculation and reporting.
  • Stable at room temperature with extended shelf life; solvent storage options at -20°C or -80°C.