Learn More
Medchemexpress LLC Ethyl octanoate | 106-32-1 | MFCD00009552 | 99.9% | 172.27 g/mol | C10H20O2 | 50 G
SDP
Catalog No. 5000302408
Encompass_Preferred
Ethyl octanoate (octanoic acid ethyl ester) is a medium-chain fatty acid ester used as a reagent, analytical standard, and flavor/fragrance component. It is a clear liquid with a fruity odor, intended for laboratory and research use in synthesis and analytical workflows.
  • High purity suitable for analytical and research applications.
  • Fruity aroma makes it a standard for flavor and fragrance testing.
  • Useful as a solvent and intermediate in organic synthesis.
  • Compatible with gas chromatography (GC) and GC-MS analysis.
  • Available in small laboratory pack sizes to support bench-scale work.
Catalog No. 50-003-02408 Supplier Medchemexpress LLC Supplier No. HYW01530550G
Ethyl octanoate (octanoic acid ethyl ester) is a medium-chain fatty acid ester used as a reagent, analytical standard, and flavor/fragrance component. It is a clear liquid with a fruity odor, intended for laboratory and research use in synthesis and analytical workflows.
  • High purity suitable for analytical and research applications.
  • Fruity aroma makes it a standard for flavor and fragrance testing.
  • Useful as a solvent and intermediate in organic synthesis.
  • Compatible with gas chromatography (GC) and GC-MS analysis.
  • Available in small laboratory pack sizes to support bench-scale work.