Learn More
Medchemexpress LLC Ethyl octanoate-d15 | 1219798-38-5 | 98.0% | 187.36 g/mol | C10H5D15O2 | 5 MG
SDP
Catalog No. 5000303260
Encompass_Preferred
Ethyl octanoate-d15 is the deuterium-labeled analogue of ethyl octanoate supplied as a stable isotope reference standard for analytical and research use. It is intended for use as an internal standard in quantitative mass spectrometry, method development, and tracer studies, offering consistent isotopic enrichment and reliable chemical purity.
  • Stable isotope internal standard for GC-MS and LC-MS
  • High chemical purity and isotopic enrichment
  • Liquid form, readily soluble in common organic solvents
  • Suitable for quantitative analysis and method validation
  • Long-term stability when stored frozen
Catalog No. 50-003-03260 Supplier Medchemexpress LLC Supplier No. HYW015305S5MG
Ethyl octanoate-d15 is the deuterium-labeled analogue of ethyl octanoate supplied as a stable isotope reference standard for analytical and research use. It is intended for use as an internal standard in quantitative mass spectrometry, method development, and tracer studies, offering consistent isotopic enrichment and reliable chemical purity.
  • Stable isotope internal standard for GC-MS and LC-MS
  • High chemical purity and isotopic enrichment
  • Liquid form, readily soluble in common organic solvents
  • Suitable for quantitative analysis and method validation
  • Long-term stability when stored frozen