Learn More
Medchemexpress LLC Ethyl octanoate-d15 | 1219798-38-5 | 98.5% | 187.36 | C10H5D15O2 | 50 MG
SDP
Catalog No. 5000303669
Encompass_Preferred
Ethyl octanoate-d15 is the deuterium-labeled analogue of ethyl octanoate provided for use as a stable-isotope tracer and internal standard in analytical chemistry. It is intended for mass spectrometry, tracer studies, and quantitative analysis where isotopic distinction from the native compound is required.
  • Deuterium-labeled (d15) ethyl octanoate for isotopic tracing.
  • High chemical purity (98.5%) suitable for analytical use.
  • Suitable as an internal standard for mass spectrometry and quantitative analysis.
  • Colorless to light yellow liquid with density 0.867 g/cm³.
  • Available in small, research-scale quantities (mg-range).
  • Certificate of analysis and safety data sheet available.
Catalog No. 50-003-03669 Supplier Medchemexpress LLC Supplier No. HYW015305S50MG
Add to Cart
Add to Cart
Ethyl octanoate-d15 is the deuterium-labeled analogue of ethyl octanoate provided for use as a stable-isotope tracer and internal standard in analytical chemistry. It is intended for mass spectrometry, tracer studies, and quantitative analysis where isotopic distinction from the native compound is required.
  • Deuterium-labeled (d15) ethyl octanoate for isotopic tracing.
  • High chemical purity (98.5%) suitable for analytical use.
  • Suitable as an internal standard for mass spectrometry and quantitative analysis.
  • Colorless to light yellow liquid with density 0.867 g/cm³.
  • Available in small, research-scale quantities (mg-range).
  • Certificate of analysis and safety data sheet available.