Learn More
Medchemexpress LLC Ethyl octanoate-d15 | 1219798-38-5 | 98.5% | 187.36 g/mol | C10H5D15O2 | 1 MG
SDP
Catalog No. 5000303606
Encompass_Preferred
Ethyl octanoate-d15 is the deuterium-labeled analogue of ethyl octanoate supplied as a high-purity liquid for use as a tracer and internal standard in quantitative NMR, GC-MS, and LC-MS analyses. It is intended for research use only and has documented storage and stability recommendations.
Catalog No. 50-003-03606 Supplier Medchemexpress LLC Supplier No. HYW015305S1MG
Ethyl octanoate-d15 is the deuterium-labeled analogue of ethyl octanoate supplied as a high-purity liquid for use as a tracer and internal standard in quantitative NMR, GC-MS, and LC-MS analyses. It is intended for research use only and has documented storage and stability recommendations.