Learn More
Medchemexpress LLC Ethyl octanoate-d15 | 1219798-38-5 | MFCD12022204 | 98.5% | 187.36 g/mol | C10H5D15O2 | 10 MG
SDP
Catalog No. 5000302479
Encompass_Preferred
Ethyl octanoate-d15 is the deuterium-labeled analogue of ethyl octanoate provided as a high-purity isotopically labeled standard for analytical and research use. It is intended for use as an internal standard or tracer in mass spectrometry and chromatographic methods to support accurate quantitation and reliable peak identification.
  • High isotopic enrichment for clear mass shifts in MS analyses.
  • Purity 98.5% supports reproducible analytical results.
  • Available in small mg-scale packages for method development.
  • Liquid form facilitates solution preparation and sample spiking.
  • Suitable for GC-MS and LC-MS internal standard applications.
Catalog No. 50-003-02479 Supplier Medchemexpress LLC Supplier No. HYW015305S10MG
Ethyl octanoate-d15 is the deuterium-labeled analogue of ethyl octanoate provided as a high-purity isotopically labeled standard for analytical and research use. It is intended for use as an internal standard or tracer in mass spectrometry and chromatographic methods to support accurate quantitation and reliable peak identification.
  • High isotopic enrichment for clear mass shifts in MS analyses.
  • Purity 98.5% supports reproducible analytical results.
  • Available in small mg-scale packages for method development.
  • Liquid form facilitates solution preparation and sample spiking.
  • Suitable for GC-MS and LC-MS internal standard applications.