Learn More
Selleck Chemical LLC Ethyl Oleate S5367-25ul
SDP
Catalog No. 503110182
Encompass_Preferred
Ethyl Oleate (Oleic acid ethyl ester) is a fatty acid ester formed by the condensation of oleic acid and ethanol usually used as a solvent for pharmaceutical drug preparations
Catalog No. 50-311-0182 Supplier Selleck Chemical LLC Supplier No. S536725UL
Ethyl Oleate (Oleic acid ethyl ester) is a fatty acid ester formed by the condensation of oleic acid and ethanol usually used as a solvent for pharmaceutical drug preparations