Learn More
Medchemexpress LLC Ethyl oleate (Standard) | 111-62-6 | 100 MG
SDP
Catalog No. 5000417644
Encompass_Preferred
Ethyl oleate (Standard) is the analytical standard of Ethyl oleate, intended for research and analytical applications. It is an orally active fatty acid ester formed from the condensation of oleic acid and ethanol. It is identified as the main fatty acid ethyl ester in the blood after alcohol ingestion. The product exhibits no obvious toxicity to rats, and its absorption, distribution, and excretion are comparable to triacylglycerol.
  • Can accelerate the drying process of certain foods.
  • Can be utilized as a liquid lipid component in nanostructured lipid carriers.
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-17644 Supplier Medchemexpress LLC Supplier No. HYN7103R100MG
Ethyl oleate (Standard) is the analytical standard of Ethyl oleate, intended for research and analytical applications. It is an orally active fatty acid ester formed from the condensation of oleic acid and ethanol. It is identified as the main fatty acid ethyl ester in the blood after alcohol ingestion. The product exhibits no obvious toxicity to rats, and its absorption, distribution, and excretion are comparable to triacylglycerol.
  • Can accelerate the drying process of certain foods.
  • Can be utilized as a liquid lipid component in nanostructured lipid carriers.
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.