Learn More
Cayman Chemical Ethyl Orsellinate, Purity: ≥95%, CAS: 2524-37-0, 500 mg
SDP
Catalog No. 502994119
Encompass_Preferred
A lichen metabolite and a derivative of lecanoric acid; inhibits the proliferation of human Hep-2 larynx carcinoma, MCF-7 breast cancer, and 786-0 kidney carcinoma cells, as well as B16-F10 murine melanoma and non-cancerous Vero cells (IC50s = 31.2, 70.3, 47.5, 64.8, and 28.1 µg/ml, respectively); toxic to brine shrimp (LC50 = 495 µM)
Catalog No. 50-299-4119 Supplier Cayman Chemical Supplier No. 22709500 MG
A lichen metabolite and a derivative of lecanoric acid; inhibits the proliferation of human Hep-2 larynx carcinoma, MCF-7 breast cancer, and 786-0 kidney carcinoma cells, as well as B16-F10 murine melanoma and non-cancerous Vero cells (IC50s = 31.2, 70.3, 47.5, 64.8, and 28.1 µg/ml, respectively); toxic to brine shrimp (LC50 = 495 µM)