Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl oxazole-4-carboxylate | 23012-14-8 | 100.0% | C6H7NO3 | 500 G
SDP

Catalog No. 5000434686
Encompass_Preferred

Ethyl oxazole-4-carboxylate is a biochemical reagent used as a biological material or organic compound for life science-related research. It is also suitable for use as a laboratory chemical and in the manufacture of substances.

  • Solid appearance
  • Color ranges from light yellow to yellow
  • Molecular weight of 141.12
  • Long-term storage stability as powder or in solvent

Catalog No. 50-004-34686 Supplier Medchemexpress LLC Supplier No. HY69111500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl oxazole-4-carboxylate is a biochemical reagent used as a biological material or organic compound for life science-related research. It is also suitable for use as a laboratory chemical and in the manufacture of substances.

  • Solid appearance
  • Color ranges from light yellow to yellow
  • Molecular weight of 141.12
  • Long-term storage stability as powder or in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.