Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl oxazole-4-carboxylate | 23012-14-8 | >99.9% | C6H7NO3 | 25 G
SDP

Catalog No. 5000359061
Encompass_Preferred

Ethyl oxazole-4-carboxylate is a biochemical reagent suitable for life science related research. It serves as a biological material or organic compound for laboratory applications and substance manufacturing. This product is for research use only and not intended for medical applications or patient use.

  • Biochemical reagent for life science research
  • High purity (>99.9%)
  • Solid appearance, light yellow to yellow color
  • Stable in powder form: store at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent: store at -80°C for 6 months or -20°C for 1 month
  • Melting point of 48°C
  • Boiling point of 202°C at 760 mmHg

Catalog No. 50-003-59061 Supplier Medchemexpress LLC Supplier No. 25GHY69111
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl oxazole-4-carboxylate is a biochemical reagent suitable for life science related research. It serves as a biological material or organic compound for laboratory applications and substance manufacturing. This product is for research use only and not intended for medical applications or patient use.

  • Biochemical reagent for life science research
  • High purity (>99.9%)
  • Solid appearance, light yellow to yellow color
  • Stable in powder form: store at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent: store at -80°C for 6 months or -20°C for 1 month
  • Melting point of 48°C
  • Boiling point of 202°C at 760 mmHg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.