Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl oxazole-4-carboxylate | 23012-14-8 | 99.96% | C6H7NO3 | 10 G
SDP

Catalog No. 5000434759
Encompass_Preferred

Ethyl oxazole-4-carboxylate is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • High purity of 99.96%
  • Solid appearance, light yellow to yellow in color
  • Molecular weight of 141.12
  • Melting point of 48°C
  • Boiling point of 202°C
  • Suitable for biochemical assay reagents

Catalog No. 50-004-34759 Supplier Medchemexpress LLC Supplier No. HY6911110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl oxazole-4-carboxylate is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • High purity of 99.96%
  • Solid appearance, light yellow to yellow in color
  • Molecular weight of 141.12
  • Melting point of 48°C
  • Boiling point of 202°C
  • Suitable for biochemical assay reagents

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.