Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl oxazole-4-carboxylate | 23012-14-8 | 99.96% | C6H7NO3 | 5 G
SDP

Catalog No. 5000433791
Encompass_Preferred

Ethyl oxazole-4-carboxylate is a biochemical reagent suitable for life science related research. It is intended for research use only and not for human or animal therapeutic or diagnostic use.

  • Molecular weight: 141.12
  • Appearance: Solid
  • Color: Light yellow to yellow
  • High purity: 99.96%

Catalog No. 50-004-33791 Supplier Medchemexpress LLC Supplier No. HY691115G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl oxazole-4-carboxylate is a biochemical reagent suitable for life science related research. It is intended for research use only and not for human or animal therapeutic or diagnostic use.

  • Molecular weight: 141.12
  • Appearance: Solid
  • Color: Light yellow to yellow
  • High purity: 99.96%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.