Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl-p-toluate, MP Biomedicals

Catalog No. ICN226435 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN226435 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN226435 Supplier MP Biomedicals, Inc Supplier No. 0522643510

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94-08-6
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
InChI Key NWPWRAWAUYIELB-UHFFFAOYSA-N
Synonym ethyl p-toluate, ethyl p-methylbenzoate, benzoic acid, 4-methyl-, ethyl ester, 4-methylbenzoic acid ethyl ester, unii-xuf0sq8l2j, p-toluic acid ethyl ester, xuf0sq8l2j, 4-methyl-benzoic acid ethyl ester, ethyl4-methylbenzoate, ethyl-p-toluate
PubChem CID 66743

Specifications

Quantity 10 g
Formula Weight 164.21
Chemical Name or Material Ethyl-P-Toluate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.