Learn More
Medchemexpress LLC Ethyl p-toluenesulfonate | 80-40-0 | MFCD00009100 | 99.9% | 200.26 g/mol | C9H12O3S | 100 G
SDP
Catalog No. 5000302246
Encompass_Preferred
Ethyl p-toluenesulfonate is a sulfonate ester used as an alkylating reagent and drug intermediate in organic synthesis. It functions as an activated ethylating agent and a good leaving group for nucleophilic substitution, and is commonly employed in the synthesis of alkyl and substituted alkyl fluorides.
  • High purity (99.93%).
  • Suitable for alkylation and nucleophilic substitution reactions.
  • Commonly used as a reagent and intermediate in pharmaceutical research.
  • Molecular formula C9H12O3S; molecular weight 200.26 g/mol.
  • Typically supplied in 100 g pack size for research use.
Catalog No. 50-003-02246 Supplier Medchemexpress LLC Supplier No. HYW014895100G
Ethyl p-toluenesulfonate is a sulfonate ester used as an alkylating reagent and drug intermediate in organic synthesis. It functions as an activated ethylating agent and a good leaving group for nucleophilic substitution, and is commonly employed in the synthesis of alkyl and substituted alkyl fluorides.
  • High purity (99.93%).
  • Suitable for alkylation and nucleophilic substitution reactions.
  • Commonly used as a reagent and intermediate in pharmaceutical research.
  • Molecular formula C9H12O3S; molecular weight 200.26 g/mol.
  • Typically supplied in 100 g pack size for research use.