Learn More
Medchemexpress LLC Ethyl palmitate | 628-97-7 | 1000 G
SDP
Catalog No. 5000421746
Encompass_Preferred
Ethyl palmitate is a CHIKV virus inhibitor with an EC50 value of 0.0068 μM. It demonstrates anti-inflammatory activity by reducing levels of TNF-α, IL-6, and NF-κB in endotoxemic rats.
  • Inhibits the expression and viral activity of CHIKV virus E2 protein
  • Decreases plasma levels of TNF-α and IL-6
  • Reduces NF-κB expression in liver and lung tissues
  • Ameliorates histopathological changes caused by LPS in a rat model
  • Inhibits phagocytic function of the reticuloendothelial system (RES)
Catalog No. 50-004-21746 Supplier Medchemexpress LLC Supplier No. HYN20861000G
Ethyl palmitate is a CHIKV virus inhibitor with an EC50 value of 0.0068 μM. It demonstrates anti-inflammatory activity by reducing levels of TNF-α, IL-6, and NF-κB in endotoxemic rats.
  • Inhibits the expression and viral activity of CHIKV virus E2 protein
  • Decreases plasma levels of TNF-α and IL-6
  • Reduces NF-κB expression in liver and lung tissues
  • Ameliorates histopathological changes caused by LPS in a rat model
  • Inhibits phagocytic function of the reticuloendothelial system (RES)