Learn More
Medchemexpress LLC Ethyl palmitate | 628-97-7 | 500 G
SDP
Catalog No. 5000420565
Encompass_Preferred
Ethyl palmitate, also known as Ethyl hexadecanoate, is a CHIKV virus inhibitor with an EC50 value of 0.0068 μM. It exhibits anti-inflammatory activity by reducing levels of TNF-α, IL-6, and NF-κB in endotoxemic rats and can ameliorate histopathological changes caused by LPS. It also inhibits the expression and viral activity of CHIKV virus E2 protein.
  • CHIKV virus inhibitor with an EC50 value of 0.0068 μM.
  • Reduces levels of TNF-α, IL-6, and NF-κB in endotoxemic rats.
  • Demonstrates anti-inflammatory activity.
  • Can ameliorate histopathological changes caused by LPS.
  • Inhibits the expression and viral activity of CHIKV virus E2 protein.
  • For research use only.
Catalog No. 50-004-20565 Supplier Medchemexpress LLC Supplier No. HYN2086500G
Add to Cart
Add to Cart
Ethyl palmitate, also known as Ethyl hexadecanoate, is a CHIKV virus inhibitor with an EC50 value of 0.0068 μM. It exhibits anti-inflammatory activity by reducing levels of TNF-α, IL-6, and NF-κB in endotoxemic rats and can ameliorate histopathological changes caused by LPS. It also inhibits the expression and viral activity of CHIKV virus E2 protein.
  • CHIKV virus inhibitor with an EC50 value of 0.0068 μM.
  • Reduces levels of TNF-α, IL-6, and NF-κB in endotoxemic rats.
  • Demonstrates anti-inflammatory activity.
  • Can ameliorate histopathological changes caused by LPS.
  • Inhibits the expression and viral activity of CHIKV virus E2 protein.
  • For research use only.