Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl palmitate, 97%

Click to view available options
Quantity:
25g
100g
500g
628-97-7
C18H36O2
284.484
MFCD00008996
XIRNKXNNONJFQO-UHFFFAOYSA-N
ethyl palmitate,hexadecanoic acid, ethyl ester,palmitic acid ethyl ester,ethyl cetylate,palmitic acid, ethyl ester,ethylpalmitate,unii-ird3m534zm,hexadecanoic acid ethyl ester,ethyl n-hexadecanoate,ird3m534zm
12366
CHEBI:84932
ethyl hexadecanoate
CCCCCCCCCCCCCCCC(=O)OCC
628-97-7
C18H36O2
284.484
MFCD00008996
XIRNKXNNONJFQO-UHFFFAOYSA-N
ethyl palmitate,hexadecanoic acid, ethyl ester,palmitic acid ethyl ester,ethyl cetylate,palmitic acid, ethyl ester,ethylpalmitate,unii-ird3m534zm,hexadecanoic acid ethyl ester,ethyl n-hexadecanoate,ird3m534zm
12366
CHEBI:84932
ethyl hexadecanoate
CCCCCCCCCCCCCCCC(=O)OCC
Applications
Ethyl palmitate acts as a lubricant on the skin’s surfaces in order to get a soft and smooth appearance. It increases cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol.

Solubility
Miscible with ethanol, dimethyl sulfoxide, chloroform and ethyl acetate. Immiscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Density 0.857
Refractive Index 1.4395
Boiling Point 192°C to 193°C (10mmHg)
Melting Point 22°C to 26°C
Flash Point 178°C (352°F)
Quantity 25g
Beilstein 1782663
Solubility Information Miscible with ethanol,dimethyl sulfoxide,chloroform and ethyl acetate. Immiscible with water.
Formula Weight 284.48
Percent Purity 97%
Chemical Name or Material Ethyl palmitate
Show More Show Less

EINECSNumber : 211-064-6

TSCA : Yes

Recommended Storage : Ambient temperatures