Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl palmitate, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014940 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4940 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4940 Supplier MilliporeSigma Supelco Supplier No. 67107250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 628-97-7
Density 0.857 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3(CH2)14 COOC2H5
MDL Number MFCD00008996
Melting Point 24°C to 26°C
Boiling Point 192°C to 193°C (10 mmHg)
Synonym Ethyl hexadecanoate; Palmitic acid ethyl ester
Type Analytical Standard
Quantity 250 mg
Formula Weight 284.48
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.