Learn More
Medchemexpress LLC Ethyl palmitate-d31 | 1215721-57-5 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN2086S
Encompass_Preferred
Ethyl palmitate-d31 is a deuterium-labeled form of Ethyl palmitate, a fatty acid ethyl ester (FAEE). It exhibits a preference for the synthesis of ethyl palmitate and ethyl oleate in human subjects following ethanol consumption. This compound serves as a hair- and skin-conditioning agent.
  • For research use only
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-20713