Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl palmitate-d31 | 1215721-57-5 | 5 MG
SDP

Catalog No. 5000420410
Encompass_Preferred

Ethyl palmitate-d31 is a deuterium labeled Ethyl palmitate. This fatty acid ethyl ester (FAEE) preferentially synthesizes ethyl palmitate and ethyl oleate over other FAEEs in human subjects after ethanol consumption. It is also used as a hair- and skin-conditioning agent. Storage is recommended at 4°C, sealed, away from moisture and light. Shipping is at room temperature in the continental US; may vary elsewhere for shipping.

  • Purity: 98.0%
  • Molecular weight: 315.67
  • Formula: C18H5D31O2
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Solubility in DMSO: 100 mg/mL (316.79 mM)

Catalog No. 50-004-20410 Supplier Medchemexpress LLC Supplier No. HYN2086S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl palmitate-d31 is a deuterium labeled Ethyl palmitate. This fatty acid ethyl ester (FAEE) preferentially synthesizes ethyl palmitate and ethyl oleate over other FAEEs in human subjects after ethanol consumption. It is also used as a hair- and skin-conditioning agent. Storage is recommended at 4°C, sealed, away from moisture and light. Shipping is at room temperature in the continental US; may vary elsewhere for shipping.

  • Purity: 98.0%
  • Molecular weight: 315.67
  • Formula: C18H5D31O2
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Solubility in DMSO: 100 mg/mL (316.79 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.