Learn More
Medchemexpress LLC Ethyl palmitate-d31 (Ethyl hexadecanoate-d31) | 1215721-57-5 | 1 MG
SDP
Catalog No. 5000422044
Encompass_Preferred
Ethyl palmitate-d31 is the deuterium-labeled form of Ethyl palmitate. Ethyl palmitate, a fatty acid ethyl ester (FAEE), shows a marked preference for the synthesis of ethyl palmitate and ethyl oleate over other FAEEs in human subjects after ethanol consumption. It is also used as a hair- and skin-conditioning agent. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs.
  • Stable heavy isotopes are used as tracers for quantitation during drug development.
Catalog No. 50-004-22044 Supplier Medchemexpress LLC Supplier No. HYN2086S1MG
Ethyl palmitate-d31 is the deuterium-labeled form of Ethyl palmitate. Ethyl palmitate, a fatty acid ethyl ester (FAEE), shows a marked preference for the synthesis of ethyl palmitate and ethyl oleate over other FAEEs in human subjects after ethanol consumption. It is also used as a hair- and skin-conditioning agent. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs.
  • Stable heavy isotopes are used as tracers for quantitation during drug development.