Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl palmitate-d31 (ethyl hexadecanoate-d31) | 1215721-57-5 | 50 MG
SDP

Catalog No. 5000421941
Encompass_Preferred

Ethyl palmitate-d31 is a deuterium-labeled form of Ethyl palmitate, a fatty acid ethyl ester (FAEE). It exhibits a preference for its own synthesis and that of ethyl oleate in human subjects following ethanol consumption. This compound is also utilized as a hair- and skin-conditioning agent, primarily for research use.

  • Deuterium labeled ethyl palmitate
  • Molecular weight 315.67
  • Formula C18H5D31O2
  • Purity 98.0%
  • Appearance is liquid
  • Color is colorless to light yellow
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Catalog No. 50-004-21941 Supplier Medchemexpress LLC Supplier No. HYN2086S50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl palmitate-d31 is a deuterium-labeled form of Ethyl palmitate, a fatty acid ethyl ester (FAEE). It exhibits a preference for its own synthesis and that of ethyl oleate in human subjects following ethanol consumption. This compound is also utilized as a hair- and skin-conditioning agent, primarily for research use.

  • Deuterium labeled ethyl palmitate
  • Molecular weight 315.67
  • Formula C18H5D31O2
  • Purity 98.0%
  • Appearance is liquid
  • Color is colorless to light yellow
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.