Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl palmitate (Standard) | 628-97-7 | 100 MG
SDP

Catalog No. 5000420562
Encompass_Preferred

Ethyl palmitate (Standard), also known as Ethyl hexadecanoate (Standard), is an analytical standard intended for research and analytical applications. It is a fatty acid ethyl ester (FAEE) that demonstrates a notable preference for its own synthesis and that of ethyl oleate over other FAEEs in human subjects following ethanol consumption. Beyond its analytical role, Ethyl palmitate is also utilized as a hair- and skin-conditioning agent.

  • Analytical standard for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS).
  • Molecular weight: 284.48; Formula: C₁₈H₃₆O₂; CAS no.: 628-97-7.
  • Appearance: Solid-liquid mixture, colorless to off-white; Purity: 98.98%.
  • Biologically relevant, synthesized in human subjects after ethanol consumption.
  • Natural sources include plants (Compositae, Umbelliferae) and microorganisms.

Catalog No. 50-004-20562 Supplier Medchemexpress LLC Supplier No. HYN2086R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ethyl palmitate (Standard), also known as Ethyl hexadecanoate (Standard), is an analytical standard intended for research and analytical applications. It is a fatty acid ethyl ester (FAEE) that demonstrates a notable preference for its own synthesis and that of ethyl oleate over other FAEEs in human subjects following ethanol consumption. Beyond its analytical role, Ethyl palmitate is also utilized as a hair- and skin-conditioning agent.

  • Analytical standard for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS).
  • Molecular weight: 284.48; Formula: C₁₈H₃₆O₂; CAS no.: 628-97-7.
  • Appearance: Solid-liquid mixture, colorless to off-white; Purity: 98.98%.
  • Biologically relevant, synthesized in human subjects after ethanol consumption.
  • Natural sources include plants (Compositae, Umbelliferae) and microorganisms.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.