Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl palmitate (Standard) | 628-97-7 | 250 MG
SDP

Catalog No. 5000420689
Encompass_Preferred

Ethyl palmitate (Standard) is an analytical standard intended for research and analytical applications. It is a fatty acid ethyl ester (FAEE) that exhibits a marked preference for the synthesis of ethyl palmitate and ethyl oleate over other FAEEs in human subjects following ethanol consumption. Additionally, it functions as a hair- and skin-conditioning agent.

  • Analytical standard for research and analytical applications
  • Fatty acid ethyl ester (FAEE)
  • Used as a hair- and skin-conditioning agent
  • Applied in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Purity of 98.98%
  • Molecular weight: 284.48
  • Chemical formula: C₁₈H₃₆O₂
  • Appearance: solid-liquid mixture, colorless to off-white
  • Initial source: Plants Compositae Dicerothamnus rhinocerotis, Plants Umbelliferae, Microorganisms

Catalog No. 50-004-20689 Supplier Medchemexpress LLC Supplier No. 250MGHYN2086R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl palmitate (Standard) is an analytical standard intended for research and analytical applications. It is a fatty acid ethyl ester (FAEE) that exhibits a marked preference for the synthesis of ethyl palmitate and ethyl oleate over other FAEEs in human subjects following ethanol consumption. Additionally, it functions as a hair- and skin-conditioning agent.

  • Analytical standard for research and analytical applications
  • Fatty acid ethyl ester (FAEE)
  • Used as a hair- and skin-conditioning agent
  • Applied in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Purity of 98.98%
  • Molecular weight: 284.48
  • Chemical formula: C₁₈H₃₆O₂
  • Appearance: solid-liquid mixture, colorless to off-white
  • Initial source: Plants Compositae Dicerothamnus rhinocerotis, Plants Umbelliferae, Microorganisms

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.