Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl palmitate (Standard) | 628-97-7 | 500 MG
SDP

Catalog No. 5000421886
Encompass_Preferred

Ethyl palmitate (Standard) is an analytical standard of Ethyl palmitate, a fatty acid ethyl ester (FAEE), intended for research and analytical applications. It is also used as a hair- and skin-conditioning agent. It is commonly utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.

  • Analytical standard of Ethyl palmitate
  • Used in research and analytical applications
  • Functions as a hair- and skin-conditioning agent
  • Suitable for HPLC, GC, and MS techniques
  • Molecular weight: 284.48
  • Formula: C₁₈H₃₆O₂
  • Solid-liquid mixture, colorless to off-white appearance
  • Can be sourced from plants and microorganisms

Catalog No. 50-004-21886 Supplier Medchemexpress LLC Supplier No. HYN2086R500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl palmitate (Standard) is an analytical standard of Ethyl palmitate, a fatty acid ethyl ester (FAEE), intended for research and analytical applications. It is also used as a hair- and skin-conditioning agent. It is commonly utilized in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.

  • Analytical standard of Ethyl palmitate
  • Used in research and analytical applications
  • Functions as a hair- and skin-conditioning agent
  • Suitable for HPLC, GC, and MS techniques
  • Molecular weight: 284.48
  • Formula: C₁₈H₃₆O₂
  • Solid-liquid mixture, colorless to off-white appearance
  • Can be sourced from plants and microorganisms

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.