Learn More
AdipoGen Ethyl pentafluoropropionate
SDP
Catalog No. 502560962
Encompass_Preferred
Chemical. CAS 426-65-3. Formula C5H5F5O2. MW 192.08. Building block for synthesis. Perfluoroalkyl carboxylate esters undergo Wittig reactions with phosphorus leading to 1-perfluoroalkyl enol ethers, which are useful precursors of various fluorinated derivatives.
Catalog No. 50-256-0962 Supplier AdipoGen Supplier No. CDXE0242G025
Chemical. CAS 426-65-3. Formula C5H5F5O2. MW 192.08. Building block for synthesis. Perfluoroalkyl carboxylate esters undergo Wittig reactions with phosphorus leading to 1-perfluoroalkyl enol ethers, which are useful precursors of various fluorinated derivatives.