Learn More
Medchemexpress LLC Ethyl phenyl(2,4,6-trimethylbenzoyl)phosphinate | 84434-11-7 | MFCD08692488 | 97.0% | 316.33 g/mol | C18H21O3P | 500 G
SDP
Catalog No. 5000300791
Encompass_Preferred
Ethyl phenyl(2,4,6-trimethylbenzoyl)phosphinate is a liquid Norrish Type I photoinitiator used to generate free radicals under UV and near-UV exposure to initiate rapid polymerization in resins, coatings, adhesives, inks, and photopolymerization-based 3D printing. It is suitable for both clear and pigmented formulations and supports two-photon induced polymerization applications.
  • Liquid form for easy incorporation into resin formulations.
  • Efficient radical generation under UV/near-UV light for fast cure.
  • Suitable for two-photon polymerization and stereolithography processes.
  • Performs well in pigmented and clear UV-curable systems for good through-cure.
  • High polymerization quantum yield for effective conversion.
Catalog No. 50-003-00791 Supplier Medchemexpress LLC Supplier No. HYW017894500G
Ethyl phenyl(2,4,6-trimethylbenzoyl)phosphinate is a liquid Norrish Type I photoinitiator used to generate free radicals under UV and near-UV exposure to initiate rapid polymerization in resins, coatings, adhesives, inks, and photopolymerization-based 3D printing. It is suitable for both clear and pigmented formulations and supports two-photon induced polymerization applications.
  • Liquid form for easy incorporation into resin formulations.
  • Efficient radical generation under UV/near-UV light for fast cure.
  • Suitable for two-photon polymerization and stereolithography processes.
  • Performs well in pigmented and clear UV-curable systems for good through-cure.
  • High polymerization quantum yield for effective conversion.