Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl phenyl(2,4,6-trimethylbenzoyl)phosphinate | 84434-11-7 | MFCD08692488 | 99.4% | 316.33 g/mol | C18H21O3P | 100 G
SDP

Catalog No. 5000301403
Encompass_Preferred

Ethyl phenyl(2,4,6-trimethylbenzoyl)phosphinate is a liquid photoinitiator used in resin 3D printing and UV/visible curing. It promotes two-photon induced polymerization and exhibits high polymerization quantum yields, making it suitable for formulations that require efficient initiation and through-cure performance.

  • Photoinitiator for two-photon induced polymerization and UV/visible curing.
  • High polymerization quantum yield for efficient initiation.
  • Purity 99.4%.
  • Molecular formula C18H21O3P and molecular weight 316.33 g/mol.
  • Suitable for resin formulation and 3D printing applications.

Catalog No. 50-003-01403 Supplier Medchemexpress LLC Supplier No. HYW017894100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl phenyl(2,4,6-trimethylbenzoyl)phosphinate is a liquid photoinitiator used in resin 3D printing and UV/visible curing. It promotes two-photon induced polymerization and exhibits high polymerization quantum yields, making it suitable for formulations that require efficient initiation and through-cure performance.

  • Photoinitiator for two-photon induced polymerization and UV/visible curing.
  • High polymerization quantum yield for efficient initiation.
  • Purity 99.4%.
  • Molecular formula C18H21O3P and molecular weight 316.33 g/mol.
  • Suitable for resin formulation and 3D printing applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.