Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethyl phenylglyoxylate | 1603-79-8 | MFCD00009120 | 99.5% | 178.19 | C10H10O3 | 5 G
SDP

Catalog No. 5000301151
Encompass_Preferred

Ethyl phenylglyoxylate (ethyl benzoylformate) is a synthetic reagent and analytical standard used as a building block and reference compound in organic synthesis and analytical workflows. It is suitable for method development, intermediate synthesis, and enzymatic studies that require a well-characterized benzoylformate derivative.

  • Purity 99.51%.
  • Chemical formula C10H10O3.
  • Molecular weight 178.19 g/mol.
  • Provided in small-scale packages suitable for research use.
  • Supplied with certificate of analysis and safety data sheet for traceability.
  • Suitable as an analytical standard and synthetic intermediate.

Catalog No. 50-003-01151 Supplier Medchemexpress LLC Supplier No. HYW0166185G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl phenylglyoxylate (ethyl benzoylformate) is a synthetic reagent and analytical standard used as a building block and reference compound in organic synthesis and analytical workflows. It is suitable for method development, intermediate synthesis, and enzymatic studies that require a well-characterized benzoylformate derivative.

  • Purity 99.51%.
  • Chemical formula C10H10O3.
  • Molecular weight 178.19 g/mol.
  • Provided in small-scale packages suitable for research use.
  • Supplied with certificate of analysis and safety data sheet for traceability.
  • Suitable as an analytical standard and synthetic intermediate.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.