Learn More
Medchemexpress LLC Ethyl phenylglyoxylate | 1603-79-8 | MFCD00009120 | 99.5% | 178.19 g/mol | C10H10O3 | 1 ML
SDP
Catalog No. 5000301150
Encompass_Preferred
Ethyl phenylglyoxylate (ethyl benzoylformate) is a synthetic reagent and analytical standard provided as a ready-to-use 10 mM solution in DMSO. It is intended for screening, assay development, enzymology studies, and use as an analytical reference. The solution format and high purity reduce preparation time and improve reproducibility in analytical workflows.
  • Ready-to-use 10 mM solution in DMSO for immediate use
  • High purity (≈99.5%) for accurate analytical results
  • Stable solution format minimizes sample preparation
  • Suitable for assay development and enzymology studies
  • Compact one mL volume for small-scale screening and reference use
Catalog No. 50-003-01150 Supplier Medchemexpress LLC Supplier No. HYW01661810MM/1ML
Ethyl phenylglyoxylate (ethyl benzoylformate) is a synthetic reagent and analytical standard provided as a ready-to-use 10 mM solution in DMSO. It is intended for screening, assay development, enzymology studies, and use as an analytical reference. The solution format and high purity reduce preparation time and improve reproducibility in analytical workflows.
  • Ready-to-use 10 mM solution in DMSO for immediate use
  • High purity (≈99.5%) for accurate analytical results
  • Stable solution format minimizes sample preparation
  • Suitable for assay development and enzymology studies
  • Compact one mL volume for small-scale screening and reference use