Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl phosphorodichloridite, 98%

Catalog No. AAB2117930
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2117930 250 g
AAB2117909 10 g
AAB2117918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2117930 Supplier Thermo Scientific Chemicals Supplier No. B2117930
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1498-42-6 | C2H5Cl2OP | 146.93 g/mol

Ethyl dichlorophosphite is used in the synthesis of peptide derivatives. It is used as an anhydride forming reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl dichlorophosphite is used in the synthesis of peptide derivatives. It is used as an anhydride forming reagent.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/moisture and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1498-42-6
Molecular Formula C2H5Cl2OP
Molecular Weight (g/mol) 146.93
MDL Number MFCD00000525
InChI Key PVCINRPAXRJLEP-UHFFFAOYSA-N
Synonym ethyl dichlorophosphite, ethyl phosphorodichloridite, ethyl dichlorophosphinite, ethylphosphorodichloridite, ethyl phosphorodichloridoite, acmc-20akws, dichloroethoxyphosphine, dichloro ethoxy phosphane, dichloro ethoxy phosphine, c2h5opcl2
PubChem CID 73905
IUPAC Name dichloro(ethoxy)phosphane
SMILES CCOP(Cl)Cl

Specifications

Density 1.284
Boiling Point 117°C to 118°C
Flash Point 4°C (39°F)
Odor Acrid
Refractive Index 1.466
Quantity 250 g
UN Number UN2924
Beilstein 1697483
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 146.94
Percent Purity 98%
Chemical Name or Material Ethyl phosphorodichloridite
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 216-098-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.