Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Phthalyl Ethyl Glycolate 95.0+%, TCI America™
SDP

Catalog No. E0164500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0164500G 500 g
E016425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0164500G Supplier TCI America Supplier No. E0164500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84-72-0
Molecular Formula C14H16O6
Molecular Weight (g/mol) 280.28
MDL Number MFCD00059377
InChI Key PZBLUWVMZMXIKZ-UHFFFAOYSA-N
Synonym Ethoxycarbonylmethyl Ethyl Phthalate, Phthalic Acid Ethoxycarbonylmethyl Ethyl Ester
PubChem CID 6785
IUPAC Name ethyl 2-[(2-ethoxy-2-oxoethoxy)carbonyl]benzoate
SMILES CCOC(=O)COC(=O)C1=CC=CC=C1C(=O)OCC

Specifications

Color Colorless
Boiling Point 320°C
Quantity 500 g
Formula Weight 280.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Phthalyl Ethyl Glycolate
EINECSNumber (3)-1313
RTECSNumber TI2060000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.