Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl picolinate, 99%

Catalog No. AC118861000
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC118861000 100 mL
AC118860250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC118861000 Supplier Thermo Scientific Chemicals Supplier No. 118861000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2524-52-9 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2524-52-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00006292
InChI Key FQYYIPZPELSLDK-UHFFFAOYSA-N
Synonym ethyl picolinate, ethyl 2-picolinate, ethyl 2-pyridinecarboxylate, picolinic acid, ethyl ester, 2-picolinic acid ethyl ester, 2-pyridinecarboxylic acid, ethyl ester, 2-ethoxycarbonyl pyridine, picolinic acid ethyl ester, 2-pyridinecarboxylic acid ethyl ester, pyridine-2-carboxylic acid ethyl ester
PubChem CID 17307
IUPAC Name ethyl pyridine-2-carboxylate
SMILES CCOC(=O)C1=CC=CC=N1

Specifications

Melting Point 2°C
Color Pink or Yellow
Density 1.1100g/mL
Boiling Point 240°C to 241°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5090 to 1.511
Quantity 100 mL
Beilstein 22, 35
Specific Gravity 1.11
Solubility Information Solubility in water: miscible
Formula Weight 151.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl picolinate, 99%
Show More Show Less
EINECSNumber 219-758-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.