Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl pipecolinate, 98+%

Catalog No. AAL1053203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1053203 1 g
AAL1053206 5 g
AAL1053214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1053203 Supplier Thermo Scientific Chemicals Supplier No. L1053203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 15862-72-3 | C8H16NO2 | 158.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15862-72-3
Molecular Formula C8H16NO2
Molecular Weight (g/mol) 158.22
MDL Number MFCD00005980
InChI Key SZIKRGHFZTYTIT-ZETCQYMHSA-O
Synonym ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720
PubChem CID 27517
IUPAC Name ethyl piperidine-2-carboxylate
SMILES CCOC(=O)[C@@H]1CCCC[NH2+]1

Specifications

Density 1.006
Boiling Point 216°C to 217°C
Flash Point 46°C (114°F)
Refractive Index 1.456
Quantity 1 g
UN Number UN1993
Beilstein 81694
Formula Weight 157.21
Percent Purity ≥98%
Chemical Name or Material Ethyl pipecolinate
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 239-990-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.