Learn More
Selleck Chemical LLC Ethyl potassium malonate
SDP
Supplier:  Selleck Chemical LLC S62221G
Encompass_Preferred
Ethyl potassium malonate (Potassium 3-ethoxy-3-oxopropanoate) is used as a competitive inhibitor of the enzyme succinate dehydrogenase It acts as a precursor to produce (trimethylsilyl)ethyl malonate which is utilized to prepare beta-ketoesters by acylation and serves as an intermediate for the preparation of ethyl tert-butyl malonate
Catalog No. 50-000-03693