Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl propiolate

Catalog No. E46607100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E46607100G 100 g
E466075G 5 g
E4660725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E46607100G Supplier Sigma Aldrich Supplier No. E46607100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Halogenated derivatives were used as substrates for direct, asymmetric alkynylation of cyclic +â-ketoesters using chiral phase-transfer catalysts
  • Also employed in a one-pot , four-component synthesis of benzene-1,2,3,5-tetracarboxylates promoted by ph3p

Specifications

CAS 623-47-2
Density 0.968 g/mL (at 25°C (literature))
Boiling Point 120°C (lit.)
Molecular Formula C5H6O2
Refractive Index n20/D 1.412 (literature)
Linear Formula HC///CCO2C2H5
MDL Number MFCD00009184
Quantity 100 g
Synonym Ethyl acetylenecarboxylate
Molecular Weight (g/mol) 98.1
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.