Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl propionylacetate, 98%

Catalog No. AC232910250
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC232910250 25 mL Glass bottle
AC232910050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC232910250 Supplier Thermo Scientific Chemicals Supplier No. 232910250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 4949-44-4 | C7H12O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4949-44-4
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009317
InChI Key UDRCONFHWYGWFI-UHFFFAOYSA-N
Synonym ethyl propionylacetate, ethyl 3-oxovalerate, ethyl 3-oxo-n-valerate, pentanoic acid, 3-oxo-, ethyl ester, 3-oxopentanoic acid ethyl ester, 3-oxo-pentanoic acid ethyl ester, ethyl 3-ketopentanoate, 3-oxovaleric acid ethyl ester, propionylacetic acid ethyl ester, ethylpropanylacetate
PubChem CID 78656
IUPAC Name ethyl 3-oxopentanoate
SMILES CCC(=O)CC(=O)OCC

Specifications

Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 83.0°C to 84.0°C (12.0 mmHg)
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4220 to 1.4240
Linear Formula C2H5COCH2CO2C2H5
Quantity 25 mL
Specific Gravity 1.01
Formula Weight 144.17
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Ethyl propionylacetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 225-593-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.