Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl pyruvate

Catalog No. E4780825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E4780825G 25 g
E47808100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E4780825G Supplier Sigma Aldrich Supplier No. E4780825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl pyruvate can undergo asymmetric henry reaction with nitromethane to form α-hydroxy α-nitro esters
  • It can be used as the model compound for α-ketoesters to study the mechanism of enantioselective hydrogenation reactions

Specifications

CAS 617-35-6
Density 1.045 g/mL (at 25°C (literature))
Boiling Point 144°C (lit.)
Molecular Formula C5H8O3
Refractive Index n20/D 1.404 (literature)
Linear Formula CH3COCOOC2H5
MDL Number MFCD00009123
Quantity 25 g
Molecular Weight (g/mol) 116.12
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.