Learn More
Medchemexpress LLC Ethyl pyruvate | 617-35-6 | MFCD00009123 | >98.0% | 1mL
SDP
Catalog No. 5000266669
Encompass_Preferred
Ethyl pyruvate is a simple derivative of the endogenous metabolite pyruvate Ethyl pyruvate is an HMGB1 release inhibitor Ethyl pyruvate can induce apoptosis by autophagy Ethyl pyruvate has anti-inflammatory antioxidant and anti-tumor activity Ethyl pyruvate can be used in the study of neurodegenerative diseases such as Alzheimer s and Parkinson s disease[1][2][3][4][5]
Catalog No. 50-002-66669 Supplier Medchemexpress LLC Supplier No. HYY136210MM/1ML
Ethyl pyruvate is a simple derivative of the endogenous metabolite pyruvate Ethyl pyruvate is an HMGB1 release inhibitor Ethyl pyruvate can induce apoptosis by autophagy Ethyl pyruvate has anti-inflammatory antioxidant and anti-tumor activity Ethyl pyruvate can be used in the study of neurodegenerative diseases such as Alzheimer s and Parkinson s disease[1][2][3][4][5]