Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl pyruvate, 98%

Catalog No. AC118945000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118945000 500 g Glass bottle
AC118941000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC118945000 Supplier Thermo Scientific Chemicals Supplier No. 118945000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 617-35-6 | C5H8O3 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-35-6
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00009123
InChI Key XXRCUYVCPSWGCC-UHFFFAOYSA-N
Synonym ethyl pyruvate, ethyl 2-oxopropionate, pyruvic acid, ethyl ester, propanoic acid, 2-oxo-, ethyl ester, pyruvic acid ethyl ester, ethyl pyroracemate, ethyl acetylformate, 2-oxo-propionic acid ethyl ester, ethyl pyruvate natural, ethyl alpha-ketopropionate
PubChem CID 12041
IUPAC Name ethyl 2-oxopropanoate
SMILES CCOC(=O)C(=O)C

Specifications

Melting Point -58.0°C
Color Yellow
Density 1.0453g/mL
Boiling Point 148.0°C to 150.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4040 to 1.4060
Linear Formula CH3COCO2C2H5
Quantity 500 g
Beilstein 03, 616
Specific Gravity 1.0453
Merck Index 15, 8135
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in ethanol,ether
Viscosity 1.18 mPa.s
Formula Weight 116.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl pyruvate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep contai
EINECSNumber 210-511-2
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.