Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (R)-(+)-1-Ethyl-2-pyrrolidinecarboxylate 99.0+%, TCI America™
SDP

Catalog No. E04695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04695G 5 g
E04691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04695G Supplier TCI America Supplier No. E04695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 381670-33-3
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
MDL Number MFCD00671533
InChI Key CUDOPASYFARLIF-UHFFFAOYNA-N
Synonym (R)-(+)-1-Ethyl-2-pyrrolidinecarboxylic Acid Ethyl Ester, (+)-1-Ethyl-D-proline Ethyl Ester, Et-D-Pro-OEt
PubChem CID 44630281
IUPAC Name ethyl 1-ethylpyrrolidine-2-carboxylate
SMILES CCOC(=O)C1CCCN1CC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 171.24
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl (R)-(+)-1-Ethyl-2-pyrrolidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.