Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (R)-(-)-3-Piperidinecarboxylate 98.0+%, TCI America™
SDP

Catalog No. N06555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06555G 5 g
N065525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06555G Supplier TCI America Supplier No. N06555G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25137-01-3
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD03093637
InChI Key XIWBSOUNZWSFKU-SSDOTTSWSA-N
Synonym r-ethyl piperidine-3-carboxylate, ethyl 3r-piperidine-3-carboxylate, ethyl r-nipecotate, r-ethyl nipecotate, r-nipecotic acid ethyl ester, r---nipecotic acid ethyl ester, r---ethyl nipecotate, r-piperidine-3-carboxylic acid ethyl ester, ethyl r---nipecotate
PubChem CID 185582
IUPAC Name ethyl (3R)-piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 157.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (R)-(-)-3-Piperidinecarboxylate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.