Learn More
Medchemexpress LLC Ethyl rosmarinate | 174591-47-0 | 100.0% | 388.37 g/mol | C20H20O8 | 10 MG
SDP
Catalog No. 5000296537
Encompass_Preferred
Ethyl rosmarinate is a phenolic compound isolated from Lycopus lucidus that exhibits antioxidative and anti-inflammatory activity. It has been reported to inhibit lung inflammation and to reduce prostaglandin E2 (PGE2) production in LPS-stimulated RAW264.7 macrophage cells (reported IC50 4.3 μM). The material is supplied as a white to off-white solid intended for research use.
  • Phenolic compound with antioxidative and anti-inflammatory activity.
  • Reported to inhibit lung inflammation.
  • Reduces PGE2 production in LPS-stimulated RAW264.7 cells (IC50 4.3 μM).
  • White to off-white solid with stated high purity.
  • Available as a 10 MG research vial.
  • Store protected from light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
Catalog No. 50-002-96537 Supplier Medchemexpress LLC Supplier No. HYN1061010MG
Add to Cart
Add to Cart
Ethyl rosmarinate is a phenolic compound isolated from Lycopus lucidus that exhibits antioxidative and anti-inflammatory activity. It has been reported to inhibit lung inflammation and to reduce prostaglandin E2 (PGE2) production in LPS-stimulated RAW264.7 macrophage cells (reported IC50 4.3 μM). The material is supplied as a white to off-white solid intended for research use.
  • Phenolic compound with antioxidative and anti-inflammatory activity.
  • Reported to inhibit lung inflammation.
  • Reduces PGE2 production in LPS-stimulated RAW264.7 cells (IC50 4.3 μM).
  • White to off-white solid with stated high purity.
  • Available as a 10 MG research vial.
  • Store protected from light; in solvent: -80°C up to 6 months, -20°C up to 1 month.