Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate, 99%

Catalog No. AC233320500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC233320500 50 g Glass bottle
AC233320100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC233320500 Supplier Thermo Scientific Chemicals Supplier No. 233320500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 7149-65-7 | C7H11NO3 | 157.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7149-65-7
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.16
MDL Number MFCD00064497
InChI Key QYJOOVQLTTVTJY-YFKPBYRVSA-N
Synonym l-pyroglutamic acid ethyl ester, ethyl s-+-2-pyrrolidone-5-carboxylate, s-ethyl 5-oxopyrrolidine-2-carboxylate, ethyl 2s-5-oxopyrrolidine-2-carboxylate, ethyl l-pyroglutamate, ethyl pidolate, h-pyr-oet, ethyl 5-oxo-l-prolinate, ethyl pyroglutamate, unii-1ps59es316
PubChem CID 2724446
IUPAC Name ethyl (2S)-5-oxopyrrolidine-2-carboxylate
SMILES CCOC(=O)C1CCC(=O)N1

Specifications

Melting Point 47°C to 51°C
Color White to Cream
Boiling Point 176°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 50 g
Specific Rotation Condition - 3.50 (20.00°C c=5,water)
Specific Rotation -3.5
Formula Weight 157.16
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-480-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.