Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl (S)-(-)-4-Chloro-3-Hydroxybutyrate

Catalog No. 4605245G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-052-45G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 46-052-45G Supplier Sigma Aldrich Supplier No. 4605245G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl (S)-4-chloro-3-hydroxybutanoate is a chiral intermediate generally used to prepare atorvastatin, a cholesterol-lowering drug.Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate can be used as a starting material in the synthesis of 4-amino-3-hydroxybutyric acid, a compound of pharmacological importance.

Specifications

CAS 90866-33-4
Density 1.19 g/mL at 25°C
Boiling Point 93°C to 95°C (5 mmHg, lit.)
Molecular Formula C6H11ClO3
Linear Formula ClCH2CH(OH)CH2CO2C2H5
Refractive Index n20/D 1.453
MDL Number MFCD00211242
Quantity 1 g
Molecular Weight (g/mol) 166.6 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.