Learn More
Medchemexpress LLC Ethyl stearate | 111-61-5 | 98.0% | 312.54 | 250 MG
SDP
Catalog No. 5000366381
Encompass_Preferred
Ethyl stearate is an ester compound produced by the reaction of ethanol and stearic acid. It is commonly used as a flavoring agent in various foods such as baked goods, beverages, and desserts. It is also used as a fragrance ingredient in the manufacture of perfume, cologne, and personal care products. Additionally, it has applications in industrial settings such as the production of lubricants and plasticizers. It is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Used as a flavoring agent in foods.
  • Used as a fragrance ingredient in personal care products.
  • Has industrial applications in lubricants and plasticizers.
  • Biochemical reagent for life science research.
Catalog No. 50-003-66381 Supplier Medchemexpress LLC Supplier No. 250MGHYW013809
Ethyl stearate is an ester compound produced by the reaction of ethanol and stearic acid. It is commonly used as a flavoring agent in various foods such as baked goods, beverages, and desserts. It is also used as a fragrance ingredient in the manufacture of perfume, cologne, and personal care products. Additionally, it has applications in industrial settings such as the production of lubricants and plasticizers. It is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Used as a flavoring agent in foods.
  • Used as a fragrance ingredient in personal care products.
  • Has industrial applications in lubricants and plasticizers.
  • Biochemical reagent for life science research.